Learn More
Across International Ai -30°C 17L Recirculating Chiller with 20L/Min Centrifugal Pump
SDP
Catalog No. 502251197
Encompass_Preferred
Ai C30 series recirculating chillers are the true thermostatic control laboratory chiller, these closed laboratory chillers are ideal for applications such as cooling rotary evaporators, analytical instrumentation, laser etching, ICP, vacuum systems, plasma etching and jacketed reactors. This system can also be used as a replacement to tap water cooling in laboratory applications.
This item is not returnable. View return policy
Catalog No. 50-225-1197 Supplier Across International Supplier No. C301710L
Add to Cart
Add to Cart
Request a Quote
Ai C30 series recirculating chillers are the true thermostatic control laboratory chiller, these closed laboratory chillers are ideal for applications such as cooling rotary evaporators, analytical instrumentation, laser etching, ICP, vacuum systems, plasma etching and jacketed reactors. This system can also be used as a replacement to tap water cooling in laboratory applications.