Learn More
Across International Ai 3x2" Electrical Heat Press w/ Dual Heating Platens -110/220V
SDP
Catalog No. 502251309
Encompass_Preferred
The Ai HydroPress 3 x 2 electrical heat press features dual built-in digital thermometers and heaters, allowing you to precisely control the temperature of your transfer. The digital timer will automatically begin a countdown as soon as the unit is pressed shut, and provides an audible alarm once the predetermined amount of time has elapsed. This is a plug and play unit and does not require an air compressor to operate.
This item is not returnable. View return policy
Catalog No. 50-225-1309 Supplier Across International Supplier No. HYDROPRESS0302
The Ai HydroPress 3 x 2 electrical heat press features dual built-in digital thermometers and heaters, allowing you to precisely control the temperature of your transfer. The digital timer will automatically begin a countdown as soon as the unit is pressed shut, and provides an audible alarm once the predetermined amount of time has elapsed. This is a plug and play unit and does not require an air compressor to operate.