Learn More
Across International Ai -40°C 1.5L Capacity Cold Trap w/ 500ml x 3 Glass Traps ETL
SDP
Catalog No. 502251299
Encompass_Preferred
The Ai 2nd Gen -40C/F T40 cold traps are ETL certified. They have a much smaller footprint, but at the same time, offering you the same cooling power.In vacuum applications, a cold trap is a device that condenses all vapors except the permanent gases into a liquid or solid. The most common objective is to prevent vapors being evacuated from an experiment from entering a vacuum pump where they would condense and contaminate it. Particularly large cold traps are necessary when removing large amounts of liquid as in freeze drying.
This item is not returnable. View return policy
Catalog No. 50-225-1299 Supplier Across International Supplier No. T40UL
The Ai 2nd Gen -40C/F T40 cold traps are ETL certified. They have a much smaller footprint, but at the same time, offering you the same cooling power.In vacuum applications, a cold trap is a device that condenses all vapors except the permanent gases into a liquid or solid. The most common objective is to prevent vapors being evacuated from an experiment from entering a vacuum pump where they would condense and contaminate it. Particularly large cold traps are necessary when removing large amounts of liquid as in freeze drying.