Learn More
Across International Ai DigiM 5L 300°C 2000 RPM PID Controlled Digital Heating Mantle
SDP
Catalog No. 502251209
Encompass_Preferred
Across International heating and stirring mantles come standard with digital display for the most accurate control. The large display interface makes it easy to quickly and precisely adjust temperature and rotation speeds. The DigiM mantle is equipped with two sets of thermocouples for proper temperature and speed detection. The heating jacket is pre-treated before shipment, which ensures that no smoke or odors (caused by heat) are released into your work environment.
This item is not returnable. View return policy
Catalog No. 50-225-1209 Supplier Across International Supplier No. DIGIM5
Across International heating and stirring mantles come standard with digital display for the most accurate control. The large display interface makes it easy to quickly and precisely adjust temperature and rotation speeds. The DigiM mantle is equipped with two sets of thermocouples for proper temperature and speed detection. The heating jacket is pre-treated before shipment, which ensures that no smoke or odors (caused by heat) are released into your work environment.