Learn More
Across International Ai EasyChill 18 Cu Ft -30°C Upright Freezer UL 110V
SDP
Catalog No. 502251395
Encompass_Preferred
The new Ai EasyChill series -30C 18 Cu Ft upright freezers is great for large storage of sample and products. Our unique compressor design with HC refrigerant offers you speedy cooling, great temperature uniformity inside the chamber, energy efficiency, and very quite operation.The E18 is UL certified and is great for storage of Covid-19 vaccines, DNA, RNA, virus samples, organs and serum.
This item is not returnable. View return policy
Catalog No. 50-225-1395 Supplier Across International Supplier No. E18.110
The new Ai EasyChill series -30C 18 Cu Ft upright freezers is great for large storage of sample and products. Our unique compressor design with HC refrigerant offers you speedy cooling, great temperature uniformity inside the chamber, energy efficiency, and very quite operation.The E18 is UL certified and is great for storage of Covid-19 vaccines, DNA, RNA, virus samples, organs and serum.