Learn More
Across International Ai EasyChill 9 Cu Ft -25°C Upright Freezer UL 115V
SDP
Catalog No. 502251393
Encompass_Preferred
The new Ai EasyChill series -25C freezers is perfect for mid to large storage of sample and products. Our unique compressor design offers you speedy cooling and great temperature uniformity inside the chamber.
This item is not returnable. View return policy
Catalog No. 50-225-1393 Supplier Across International Supplier No. E09.110
The new Ai EasyChill series -25C freezers is perfect for mid to large storage of sample and products. Our unique compressor design offers you speedy cooling and great temperature uniformity inside the chamber.