Learn More
Across International Ai EasyVac 7 cfm Vacuum Pump with Oil Mist Filter ETL/CE
SDP
Catalog No. 502251211
Encompass_Preferred
Ai EasyVac 7 single-stage sliding-vane rotary compact vacuum pump with 3/8" hose barb vacuum connector. Comes with a 3-foot food grade silicone hose and a exhaust oil mist filter which also returns trapped pump oil back to the pump and makes the pump quieter to run. It is important to note that pumps should always be ran in a well ventilated area.
This item is not returnable. View return policy
Catalog No. 50-225-1211 Supplier Across International Supplier No. EV7
Ai EasyVac 7 single-stage sliding-vane rotary compact vacuum pump with 3/8" hose barb vacuum connector. Comes with a 3-foot food grade silicone hose and a exhaust oil mist filter which also returns trapped pump oil back to the pump and makes the pump quieter to run. It is important to note that pumps should always be ran in a well ventilated area.