Learn More
Across International Ai Glacier 21 Cu Ft -86C Ultra Low Temp Chest Freezer ETL 220V
SDP
Catalog No. 502251356
Encompass_Preferred
The all new Ai Glacier 21 Cu Ft -86C ultra low Temprature chest freezer is perfect for extra-large storage of sample and products. Our unique dual parallel compressor design offers you speedy cooling and great temperature uniformity inside the chamber.
This item is not returnable. View return policy
Catalog No. 50-225-1356 Supplier Across International Supplier No. G21C
Add to Cart
Add to Cart
Request a Quote
The all new Ai Glacier 21 Cu Ft -86C ultra low Temprature chest freezer is perfect for extra-large storage of sample and products. Our unique dual parallel compressor design offers you speedy cooling and great temperature uniformity inside the chamber.