Learn More
Across International Ai RapidChill 4 CF -86°C Stackable Ultra low Freezer UL 110V
SDP
Catalog No. 502251360
Encompass_Preferred
The Ai RapidChill series -86C ultra-low upright ULT freezers is designed for fast cooling, ultra-low power consumption, very low noise, and ease of maintenance. All RapidChill ultra-low freezers are UL certified.
This item is not returnable. View return policy
Catalog No. 50-225-1360 Supplier Across International Supplier No. G04H
Add to Cart
Add to Cart
Request a Quote
The Ai RapidChill series -86C ultra-low upright ULT freezers is designed for fast cooling, ultra-low power consumption, very low noise, and ease of maintenance. All RapidChill ultra-low freezers are UL certified.