Learn More
Across International Ai SolventVap 2L Rotary Evaporator with Electric Flask Lift 110V
SDP
Catalog No. 502251288
Encompass_Preferred
Ai SolventVap 2L (evaporating flask capacity) series rotary evaporators come with a modular heating bath, vertical glassware, and digital temperature/rotation control. Its optimized bath volume allows for rapid heat-up times. The condenser unit has specially designed glass tubes that utilize surface area extremely efficiently. Thats why our SolventVap evaporators consistently deliver reliable and reproducible process results that are usually reserved for more costly systems. Another important aspect of the SolventVap is the safety features, which includes dry-run protection, water bath over-temperature shut-off protection, and fuse protection.
This item is not returnable. View return policy
Catalog No. 50-225-1288 Supplier Across International Supplier No. SE05X.110
Ai SolventVap 2L (evaporating flask capacity) series rotary evaporators come with a modular heating bath, vertical glassware, and digital temperature/rotation control. Its optimized bath volume allows for rapid heat-up times. The condenser unit has specially designed glass tubes that utilize surface area extremely efficiently. Thats why our SolventVap evaporators consistently deliver reliable and reproducible process results that are usually reserved for more costly systems. Another important aspect of the SolventVap is the safety features, which includes dry-run protection, water bath over-temperature shut-off protection, and fuse protection.