Learn More
Across International Ai SST -20°C to 99°C 7L Compact Recirculating Chiller
SDP
Supplier:  Across International C207SST
Encompass_Preferred
Ai C20 series recirculating chillers are the true thermostatic control laboratory chiller, this closed laboratory chiller is ideal for applications such as cooling rotary evaporators, analytical instrumentation, laser etching, ICP, vacuum systems, plasma etching and jacketed reactors. This system can also be used as a replacement to tap water cooling in laboratory applications. Ai C20 are compressor-based recirculating chillers that offer outstanding performance and high reliability as well as quiet operation and ease-of-use. Powerful force/suction pump provides constant flow rates, while 2nd generation controller gives you the ideal solution when you need precise temperature control or cooling below ambient temperature.
Catalog No. 50-225-1195