Learn More
Cayman Chemical AICA Ribonucleotide, Purity: ≥95%, CAS: 3031-94-5, 5 mg
SDP
Catalog No. 502948092
Encompass_Preferred
An AMPK activator; activates AMPK in a cell-free assay from 1-500 µM; inhibits lipolysis and lipogenesis in primary rat adipocytes from 100-500 µM; increases insulin sensitivity and prevents pancreatic β-cell damage in Zucker diabetic fatty rats at 0.5 mg/kg; induces cardiac mobilization and myofibroblast differentiation of endogenous CD44+CD45-/- MSCs, as well as decreases cardiac scar formation, in an aged mouse model of LAD occlusion-induced myocardial infarction at 0.5 mg/kg
Catalog No. 50-294-8092 Supplier Cayman Chemical Supplier No. 339075 MG
An AMPK activator; activates AMPK in a cell-free assay from 1-500 µM; inhibits lipolysis and lipogenesis in primary rat adipocytes from 100-500 µM; increases insulin sensitivity and prevents pancreatic β-cell damage in Zucker diabetic fatty rats at 0.5 mg/kg; induces cardiac mobilization and myofibroblast differentiation of endogenous CD44+CD45-/- MSCs, as well as decreases cardiac scar formation, in an aged mouse model of LAD occlusion-induced myocardial infarction at 0.5 mg/kg