Learn More
Medchemexpress LLC AICA-riboside, 5′-phosphate | 3031-94-5 | 100.0% | 338.21 g·mol⁻¹ | C9H15N4O8P | 5 MG
SDP
Catalog No. 5000222444
Encompass_Preferred
AICA-riboside, 5′-phosphate is the 5′-phosphorylated form of AICA-riboside that acts as an AMP mimetic and modulates adenosine uptake. It is used as a research reagent to activate AMP-activated protein kinase (AMPK) and to investigate cellular energy metabolism, metabolic disease models, and neuroprotective mechanisms.
  • Activates AMP-activated protein kinase via AMP mimetic activity.
  • Modulates adenosine uptake and extracellular adenosine concentration.
  • Used in studies of metabolic and neurological disorders.
  • Provided as a solid with manufacturer-stated high purity.
  • Available in small pack sizes suitable for laboratory research.
Catalog No. 50-002-22444 Supplier Medchemexpress LLC Supplier No. HY1343565MG
AICA-riboside, 5′-phosphate is the 5′-phosphorylated form of AICA-riboside that acts as an AMP mimetic and modulates adenosine uptake. It is used as a research reagent to activate AMP-activated protein kinase (AMPK) and to investigate cellular energy metabolism, metabolic disease models, and neuroprotective mechanisms.
  • Activates AMP-activated protein kinase via AMP mimetic activity.
  • Modulates adenosine uptake and extracellular adenosine concentration.
  • Used in studies of metabolic and neurological disorders.
  • Provided as a solid with manufacturer-stated high purity.
  • Available in small pack sizes suitable for laboratory research.