Learn More
Medchemexpress LLC HY-13417 100mg , AICAR CAS:2627-69-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13417100MG
Encompass_Preferred
HY-13417 100mg AICAR CAS: 2627-69-2 AICAR (Acadesine) is an adenosine analog and a AMPK activator. AICAR regulates the glucose and lipid metabolism, and inhibits proinflammatory cytokines and iNOS production. AICAR is also an autophagy, YAP and mitophagy inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0059