Learn More
AdipoGen AICAR (10 mg)
SDP
Supplier:  AdipoGen AGCR10061M010
Encompass_Preferred
Chemical. CAS: 2627-69-2. Formula: C9H14N4O5. MW: 258.2. Cell permeable AMP-activated protein kinase (AMPK) activator. Insulin mimetic. Adipocyte differentiation inhibitor. Apoptosis inducer. PPARalpha inhibitor. mTOR inhibitor. P70S6K inhibitor. LPS-induced TNF-alpha production inhibitor. TORC2 phosphorylation inducer. Anti-inflammatory. Anti-tumor compound. Autophagy inhibitor. HSP90 inhibitor. Autophagy inducer
Catalog No. 50-205-3815