Learn More
Apexbio Technology LLC AICAR, 2627-69-2, MFCD00869751, 200 mg
SDP
Catalog No. 501011662
Encompass_Preferred
MF: C9H14N4O5, MW: 258.23, Solubility: 12.9 mg/mL in DMSO, 52.9 mg/mL in H2O,insoluble in EtOH, Chemical Name: 5-amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl)imidazole-4-carboxamide. AICAR is a cell-permeable, allosteric activator of AMP-activated protein kinase (AMPK).
Catalog No. 50-101-1662 Supplier Apexbio Technology LLC Supplier No. A8184200
MF: C9H14N4O5, MW: 258.23, Solubility: 12.9 mg/mL in DMSO, 52.9 mg/mL in H2O,insoluble in EtOH, Chemical Name: 5-amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl)imidazole-4-carboxamide. AICAR is a cell-permeable, allosteric activator of AMP-activated protein kinase (AMPK).