Learn More
Abcam AICAR (Acadesine/AICA riboside), AMPK activator, 250MG
Catalog No. 502584618
Encompass_Preferred
MW 258.23 Da, Purity >98%. Cell-permeable activator of AMP-activated protein kinase. Is taken up into cells by adenosine transporters and phosphorylated by adenosine kinase to the active nucleotide ZMP (5-aminoimidazole-4-carboxamide ribonucleoside), which mimics effects of AMP on the AMPK system. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4618 Supplier Abcam Supplier No. AB120358250MG
MW 258.23 Da, Purity >98%. Cell-permeable activator of AMP-activated protein kinase. Is taken up into cells by adenosine transporters and phosphorylated by adenosine kinase to the active nucleotide ZMP (5-aminoimidazole-4-carboxamide ribonucleoside), which mimics effects of AMP on the AMPK system. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement