Learn More
Apexbio Technology LLC AICAR phosphate 681006-28-0 50mg
SDP
Catalog No. 5000088595
Encompass_Preferred
AICAR phosphate (CAS 681006-28-0) also known as Acadesine is an activator of AMP-activated protein kinase (AMPK) In studies involving B-cell chronic lymphocytic leukemia (B-CLL) cells AICAR phosphate induces apoptosis in a dose-dependent manner with an EC50 of approximately 380 60 M This apoptotic effect is associated with caspase activation and mitochondrial cytochrome c release Notably AICAR phosphate at concentrations of 2 4 mM has minimal impact on T lymphocyte viability from B-CLL patients indicating selective activity These properties make it a valuable tool for research on AMPK signaling and B-cell malignancy mechanisms
Catalog No. 50-000-88595 Supplier Apexbio Technology LLC Supplier No. B121150
AICAR phosphate (CAS 681006-28-0) also known as Acadesine is an activator of AMP-activated protein kinase (AMPK) In studies involving B-cell chronic lymphocytic leukemia (B-CLL) cells AICAR phosphate induces apoptosis in a dose-dependent manner with an EC50 of approximately 380 60 M This apoptotic effect is associated with caspase activation and mitochondrial cytochrome c release Notably AICAR phosphate at concentrations of 2 4 mM has minimal impact on T lymphocyte viability from B-CLL patients indicating selective activity These properties make it a valuable tool for research on AMPK signaling and B-cell malignancy mechanisms