Learn More
Enzo Life Sciences AIDA (25mg). CAS: 168560-79-0
SDP
Supplier:  Enzo Life Sciences ALX550101M025
Encompass_Preferred
Potent and selective ligand for group I mGluRs. Shows no effect on group II or group III mGluRs. Has no effect on ionotropic glutamate receptors. Alternative name: (+/-)-1-Aminoindane-1,5-dicarboxylic acid, UPF 523. Purity: ≥96%. Solubility: Soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9482