Learn More
Aviva Systems Biology Corp AIDA Antibody - N-terminal region (ARP63070_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63070_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AIDA. It was validated on Western Blot. . Alternate names: C1orf80, FLJ12806, FLJ32421, RP11-378J18.7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-863