Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ AIDS Kit I: Simulation of HIV-1 Detection
SDPSCI_ED

Catalog No. s11271 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S11271 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11271 Supplier Edvotek™ Supplier No. 271
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Complete instructions, background information, and study questions; biologicals and reagents; three microtiter plates; 50 transfer pipettes; 60 microtest tubes with attached caps; twenty-five 1mL pipettes; two 50mL plastic tubes.

Requires

Requirements:

  • Distilled or deionized water
  • Beakers z 37°C incubation oven
  • Automatic micro- pipettes and tips (optional)

In this experiment, students will perform an ELISA test by coating microtiter plate wells with simulated HIV antigen and then test simulated donor serum for anti-HIV antibodies.

A model for HIV-1 detection without the use of blood

Simulates the screening for antibodies against human immunodeficiency virus (HIV-1). Enzyme-linked immunoabsorbent assay (ELISA) is currently used for initial clinical screening of the blood supply for HIV-1. Students add simulated HIV antigen to wells in the microtiter plate and follow it with washing and introduction of simulated antibodies of HIV-1 positive or negative samples. A second antibody is added which binds to the first antibody of HIV-1 positive samples and results in a color change.

Note: some kit contents may need to be redeemed by mail.

Order Info

Kit ships with plasticware and instructions on how to obtain remaining materials directly from the supplier, Edvotek.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Detects hiv infection indirectly using an elisa test against hiv antibodies in the blood
Includes Instructions, simulated hiv antigens, serum samples, antibodies, buffers, microtiter plates, assorted pipets and microtest tubes
Time Range 60 min.
Content And Storage Refrigerated
Class Size 10 Lab Groups
Product Type AIDS Kit I
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.