Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ AIDS Kit III: Simulation of HIV Detection by Protein Electrophoresis Experiment Kit
SDPSCI_ED

Catalog No. S29013 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
6 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S29013 6 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S29013 Supplier Edvotek™ Supplier No. 151
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

In this experiment, students will use SDS-PAGE to simulate the identification of HIV proteins in simulated patient samples. Edvotek™ AIDS Kit III: Simulation of HIV Detection by Protein Electrophoresis Experiment Kit test results are used to diagnose an HIV infection.

  • For 6 Lab Groups
  • Electrophoresis (60 min) Staining/Destaining (Optional, 2 hrs)
  • Level: Intermediate

Specifications

Grade 9 to Undergraduate
Includes instructions, denatured LyphoProtein™ samples, standard protein markers, practice gel loading solution, buffer, transfer pipettes, Protein InstaStain™
Time Range Electrophoresis (60 min) Staining/Destaining (Optional, 2 hrs)
Content And Storage Some components require storage -20 ̊C. 6 Months.
Class Size 6 Lab Groups
Product Type AIDS Kit III: Simulation of HIV Detection by Protein Electrophoresis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.