Learn More
Cayman Chemical AIF Blockng PeptIde 1ea
SDP
Catalog No. 502912644
Encompass_Preferred
A peptide fragment of ApoE and an a7 nAChR antagonist (IC50 720 nM) suppresses TNF-a and NO release in BV-2 microglia from 10-50 UM reduces LPS-induced increases in brain levels of TNF-a and IL-6 in mice decreases hippocampal neuronal degeneration and the latency to find the platform in the Morris water maze in a mouse model of closed head injury induced by surgical midline impact at 203 and 406 Ug/kg delays disease onset and reduces disease severity in a mouse model of EAE at 1 mg/kg
Catalog No. 50-291-2644 Supplier Cayman Chemical Supplier No. 3607731 EA
A peptide fragment of ApoE and an a7 nAChR antagonist (IC50 720 nM) suppresses TNF-a and NO release in BV-2 microglia from 10-50 UM reduces LPS-induced increases in brain levels of TNF-a and IL-6 in mice decreases hippocampal neuronal degeneration and the latency to find the platform in the Morris water maze in a mouse model of closed head injury induced by surgical midline impact at 203 and 406 Ug/kg delays disease onset and reduces disease severity in a mouse model of EAE at 1 mg/kg