Learn More
Aviva Systems Biology Corp AIFM1 Antibody - middle region (AVARP02013_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP02013_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AIFM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AIFM1. Alternate names: AIF, MGC111425, PDCD8, COXPD6ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62477