Learn More
Aviva Systems Biology Corp AIFM2 Antibody - middle region (AVARP09054_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP09054_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AIFM2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AIFM2. Alternate names: AMID, PRG3, RP11-367H5.2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62590