Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci AIFM2 (CT) Peptide
SDP

Catalog No. 16595427
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-427 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-427 Supplier ProSci Supplier No. 9653P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

AIFM2 (CT) Peptideis used for blocking the activity of theAIFM2 (CT) Antibody

AIFM2 Antibody: AIFM2 blocks ferroptosis independent of ubiquinol metabolism. AIFM2 is a key component of a non-mitochondrial CoQ antioxidant system that acts in parallel to the canonical glutathione-based GPX4 pathway. AIFM2 confers protection against ferroptosis elicited by GPX4 deletion. AIF-2 was found to be upregulated in human glioma tissues and cell lines. Downregulation of AIF-2 reduced cell proliferation and induced G1 cell cycle arrest. AIF and its family member protein, AMID, are rotenone-sensitive NADH:ubiquinone oxidoreductases (of the NDH-2 type). AIF-M2 lessens survival cell signaling in the presence of foreign (e.g. bacterial and (retro)viral) cytosolic DNA, thus contributing to the onset of apoptosis

Specifications

Gene ID (Entrez) 84883
Species Human
Accession Number Q9BRQ8
Common Name apoptosis inducing factor mitochondria associated
Concentration 200 μg/mL
Content And Storage Store AIFM2 (CT) Peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol AIMF2
Quantity 0.05 mg
Sequence 12 amino acids near the carboxy terminus of AIFM2 (CT).
Synonym AIMF2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.