Learn More
Aviva Systems Biology Corp AIFM3 Antibody - middle region (ARP52745_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52745_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AIFM3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AIFM3. Alternate names: AIFL, FLJ30473ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60817