Learn More
TARGETMOL CHEMICALS INC AILANTHONE 10MG
SDP
Catalog No. 502576152
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ailanthone ((delta)13-Dehydrochaparrinone) is an effective inhibitor of both full-length androgen receptor (AR) (IC50 69 nM) and constitutively active truncated AR splice variants (IC50 309 nM). purity: 99%
Catalog No. 50-257-6152 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ020910MG
Also available in 1 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ailanthone ((delta)13-Dehydrochaparrinone) is an effective inhibitor of both full-length androgen receptor (AR) (IC50 69 nM) and constitutively active truncated AR splice variants (IC50 309 nM). purity: 99%