Learn More
Medchemexpress LLC HY-N1943 5mg Medchemexpress, Ailanthone CAS:981-15-7 Purity:>98%
SDP
Catalog No. 501933863
Encompass_Preferred
Medchemexpress, HY-N1943 5mg Ailanthone CAS:981-15-7 Ailanthone (Δ13-Dehydrochaparrinone) is a potent inhibitor of both full-length androgen receptor (AR) ( IC 50 =69 nM) and constitutively active truncated AR splice variants (AR 1-651 IC 50 =309 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3863 Supplier Medchemexpress LLC Supplier No. HYN19435MG
Medchemexpress, HY-N1943 5mg Ailanthone CAS:981-15-7 Ailanthone (Δ13-Dehydrochaparrinone) is a potent inhibitor of both full-length androgen receptor (AR) ( IC 50 =69 nM) and constitutively active truncated AR splice variants (AR 1-651 IC 50 =309 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.