Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories Aim™ ASO Test

Catalog No. 23111306
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-306 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-306 Supplier Germaine™ Laboratories Supplier No. 31050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Germaine™ Laboratories Aim™ ASO Test is a latex agglutination test for the qualitative and semi-quantitative determination of ASO (Antistreptolysin-O) in non-diluted serum

  • Two-minute slide test
  • No initial dilution of patient samples is required for this test
  • ASO latex reagent consists of latex particles coated with streptolysin-O

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes Anti-Streptolysin O (ASO)
Sample Type Non-Diluted Serum
Disposable Single-use
Quantity 50 Tests
Test Time 2 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.