Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories Aim™ CRP Test

Catalog No. 23111304
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-304 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-304 Supplier Germaine™ Laboratories Supplier No. 32100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Germaine™ Laboratories, Inc. Aim™ CRP Test is a latex agglutination test for the qualitative and semi-quantitative determination of C-Reactive Protein (CRP) in non-diluted serum

  • Two-minute slide test
  • No initial dilution required
  • Two liquid controls included
Note:  Includes 1 reaction card (with 6 ovals). For information on purchasing additional reaction cards, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes CRP
Sample Type Non-Diluted Serum
Disposable Single-use
Includes Reagents: CRP Latex, Positive control, Neg. control, Glycine Saline Buffer; 100 pipettes; One reaction card.
Quantity 100 Tests
Test Time 2 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.