Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories Aim™ RF Test

Catalog No. 23111305
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-305 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-305 Supplier Germaine™ Laboratories Supplier No. 35100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Germaine™ Laboratories Aim™ RF Test is a latex agglutination test for the qualitative and semi-quantitative determination of Rheumatoid Factor (RF) in non-diluted serum

  • Two-minute slide test
  • No initial dilution required
  • Two liquid controls included

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes Rheumatoid Factor (RF)
Sample Type Non-Diluted Serum
Disposable Single-use
Quantity 100 Tests
Test Time 2 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.