Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ AIM V™ Medium, liquid (research grade)
GREENER_CHOICE

Catalog No. 12055083
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-055-083 1000 mL
12-055-091 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-055-083 Supplier Gibco™ Supplier No. 12055083
  • $211.00
    $185.65 / Each of 1

    Save $25.35

    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

AIM V (research grade) is a medium designed for the proliferation and/or manipulation of T cells, dendritic cells, and other immune cells.

AIM V (research grade) is a medium designed for the proliferation and/or manipulation of T cells, dendritic cells, and other immune cells.

AIM V Medium contains L-glutamine, streptomycin sulfate at 50 μg/mL, and gentamicin sulfate at 10 μg/mL.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Monocytes, Dendritic Cells, T Cells, Hybridomas, PBMCs, Fibroblasts, Macrophages, Myeloma Cells, Lymphocytes
Form Liquid
Product Type Serum Free Medium (SFM)
With Additives Glutamine, Phenol Red, HSA
Product Line AIM V
Quantity 1000 mL
Shipping Condition Room Temperature
Included Antibiotics Streptomycin Sulfate, Gentamicin
Purity or Quality Grade Research Grade
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Mammalian Cell Culture
Show More Show Less
What is the difference between AIM V Medium (Cat. Nos. 12055083, 12055091) and CTS AIM V SFM (Cat. Nos. 0870112DK, 0870112BK)?

The difference is in the intended usage of the two media. AIM V Medium (Cat. Nos. 12055083, 12055091) is for research use only whereas CTS AIM V SFM (Cat. Nos. 0870112DK, 0870112BK) is for research use or manufacturing of cell, gene, or tissue-based products.

Which serum-free medium would you recommend for culture and expansion of mouse T cells?

You can try using AIM V Medium (Cat. No. 12055083 or 12055091) for growth of mouse T cells. For mouse cells, we recommend adding 50 µM beta-mercaptoethanol (Cat. No. 21985023).

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.