Learn More
Aviva Systems Biology Corp AIM2 Antibody - N-terminal region (ARP50505_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50505_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AIM2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AIM2. Alternate names: PYHIN4ApplicationWB. Species reactivity: Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-53633