Learn More
Medchemexpress LLC AIM2 Human sf9 GS 50ug
SDP
Catalog No. 5000181517
Encompass_Preferred
AIM2 Protein a member of the IFI20X/IFI16 family exhibits potential regulatory functions in cell proliferation and tumorigenic reversion Induced by interferon-gamma AIM2 plays a key role in immune responses Notably expressed in tissues like lymph node and appendix AIM2 is a significant player in diverse cellular processes and immune-related functions AIM2 Protein Human (sf9 GST) is the recombinant human-derived AIM2 protein expressed by Sf9 insect cells with N-GST labeled tag
Catalog No. 50-001-81517 Supplier Medchemexpress LLC Supplier No. HYP7281650UG
AIM2 Protein a member of the IFI20X/IFI16 family exhibits potential regulatory functions in cell proliferation and tumorigenic reversion Induced by interferon-gamma AIM2 plays a key role in immune responses Notably expressed in tissues like lymph node and appendix AIM2 is a significant player in diverse cellular processes and immune-related functions AIM2 Protein Human (sf9 GST) is the recombinant human-derived AIM2 protein expressed by Sf9 insect cells with N-GST labeled tag