Learn More
Aviva Systems Biology Corp AIMP2 Antibody - N-terminal region (ARP59346_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59346_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AIMP2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human AIMP2. Alternate names: JTV-1, JTV1, P38, PRO0992ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-286-670