Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIMS™ ATS-3 General Rodent Tattoo System Pigment
GSA_VA

Catalog No. p-4278522
Change view
Click to view available options
Color:
Black
Green
Red
Capacity (English):
0.16 oz.
2 oz.
Capacity (Metric):
5 mL
59.14 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Capacity (English) Capacity (Metric)
14-370-167 Black 2 oz. 59.14 mL
14-370-168 Black 0.16 oz. 5 mL
14-370-169 Red 0.16 oz. 5 mL
14-370-170 Green 0.16 oz. 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-370-167 Supplier AIMS™ Supplier No. 2420
Only null left
Add to Cart
Edge
Add to Cart

Will not alter the physiology of animals or contaminate other lab animals in the facility

  • Safe
  • Nontoxic

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Capacity (English) 2 oz.
Capacity (Metric) 59.14 mL
Color Black
For Use With (Application) In-vivo and in-vitro Toxicology Assay
Type Tattoo Pigment
For Use With (Equipment) ATS-3 general rodent tattoo system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.