Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIMS™ Body-Contour Cone-Shaped Plastic Disposable Rodent Restraints

Catalog No. 14370118
Change view
Click to view available options
Color:
Black
Clear
Rose
Size:
Small
Medium
Large
Weight Range:
≤120 g
≤225 g
≤450 g
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size Weight Range
14-370-118 Black Large ≤450 g
14-370-110 Clear Small ≤120 g
14-370-111 Clear Medium ≤225 g
14-370-112 Clear Large ≤450 g
14-370-116 Black Small ≤120 g
14-370-114 Rose Medium ≤225 g
14-370-115 Rose Large ≤450 g
14-370-113 Rose Small ≤120 g
14-370-117 Black Medium ≤225 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 14-370-118 Supplier AIMS™ Supplier No. 1803BV
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Instructions and twist ties

Humane, safe, disposable rodent restraint system

Available in a variety of sizes and colors. Cone-shaped bags provide easier animal capture and increased animal comfort.

  • Precut sizes for quick use
  • Easy-Flex-PETM provides effortless wrapping and added animal comfort
  • Body-Contour cone shape provides easier restraint, exceptional comfort, and reduced animal stress
  • Black and rose color bags reduce natural light and aids in reducing animal agitation and stress

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.

Specifications

Color Black
For Use With (Application) Tail Tattooing, Tail Vein Injections, Oral Dosing, Decapitation and General Restraint
Type Rodent Restraint
Disposable Single-use
Includes Instructions, twist ties
Material Plastic
Shape Cone
Size Large
Weight Range ≤450 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.