Learn More
AIMS™ CO2 Flowmeters: Model LC500
Catalog No. 14370201
Change view
Click to view available options
Model:
LC500
Catalog No. Model
14-370-201 LC500
1 options
Catalog No. 14-370-201 Supplier AIMS™ Supplier No. LC500
Add to Cart
Add to Cart
Calibrated flowmeter that accurately delivers CO2 to chamber
  • Three-way ball valve offers quick chamber fill when needed
  • Allows lab personnel to meet all requirements of new 2013 AVMA Guidelines for CO2 euthanasia
  • Enclosure: 12.7W x 16.5cm H (5 x 6.5 in.)
CO2 flow rate: 1 to 10L/minute
Specifications
Model LC500