Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIMS™ Tattoo Needle Tip Inspection Scope

Catalog No. 14370149
Change view
Click to view available options
Color:
Black/White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-370-149 Black/White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-370-149 Supplier AIMS™ Supplier No. 4000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quickly Scan Tattoo Needle Tips For Damage

 

    • Used to quickly inspect tattoo needle tips for damage
    • Lighted magnifier allows to easy inspection of tattoo needles for defects

 

Specifications

Color Black/White
Type Tattoo Needle Tip Inspection Scope
For Use With (Equipment) NEO-9, ATS-3, ATS-3L Tattoo Systems
Includes Battery
Dimensions (L x W) 3.25 x 1.75 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.