Learn More
AIMS™ Tattoo Machine Fine Tip Needle Tube
Catalog No. 14370127
Change view
Click to view available options
For Use With (Application):
Lab Animal Tattoo Identification
Catalog No. For Use With (Application)
14-370-127 Lab Animal Tattoo Identification
1 options
Catalog No. 14-370-127 Supplier AIMS™ Supplier No. 1701
Add to Cart
Add to Cart
Fine Tip Needle Tube Used To Tattoo Neonate Rodents
  • For tattooing neonate rodents
  • Calibrated for easy and accurate needle adjustment
Specifications
For Use With (Application) Lab Animal Tattoo Identification
Type Needle Tube
For Use With (Equipment) NEO-9, ATS-3, ATS-3L Tattoo Systems
Material Stainless Steel