Learn More
AIMS™ NEO-9 Neonate Rodent Tattooing System
Catalog No. 14370129
Change view
Click to view available options
Certifications:
AIMS
Catalog No. Certifications
14-370-129 AIMS
1 options
Catalog No. 14-370-129 Supplier AIMS™ Supplier No. NEO9
Add to Cart
Add to Cart
Request a Quote
Neonate tattooing made simple, quick and easy
For tattooing P0-P15 neonate rodents of any color/strain. Comprehensive tattoo ID kit comes with everything you need to tattoo appoximately 2000 neonates. Training guide provides step-by-step guidance with detailed illustrations of ID schemes with small and large number ranges. Neonate tattoo identification is a practical and humane alternative to toe clipping. Toe tattoos allow you to use your existing numbering system for toe clipping without removing the toe. Heavy duty tattoo equipment is built for the laboratory environment.
  • Tattoo P0-P15 mice or rats; can be used for any color neonate
  • No anesthesia needed
  • Tattoo markings are permanent
  • Reduce animal stress; quick and safe
  • Economical; costs aprox. 2 cents/neonate
  • Comprehensive manual with DVD instruction
  • Multiple ID schemes available
  • Unique numbers up to 10,000
  • Custom micro tattooing platform
  • Micro controller with HDUI power supply
  • Foot pedal ON/OFF; preset speeds
  • Ultra lightweight machine
  • Non-skid machine platform with pigment holder and needle scanning area
  • Micro tattooing station; light, skin prep holder, 4X magnification
  • AIMS certified-safe tattoo pigment
  • AIMS skin prep (reduces fading)
  • Standard/fine tip tattoo needles
  • Finger guards; needle scanning scope
  • Ideal for managing transgenic animals and geotyping
  • Unlimited technical support; on-site training available
  • Carrying case
Warranty and Services
1 year
Specifications
For Use With (Application) Managing Transgenic Animals and Genotyping
Certifications AIMS
Includes Tattoo markings permanent, instructions with DVD, unique numbers, custom micro tattooing platform, unlimited technical support, power supply, ultra lightweight machine, non skid machine platform, micro tattooing station, tattoo pigment, tattoo needles