Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIMS™ Permanent Tattoo Pigment for NEO-9 Neonate Tattoo System
GSA_VA

Catalog No. 14370171
Change view
Click to view available options
Capacity (English):
0.5 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
14-370-171 0.5 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-370-171 Supplier AIMS™ Supplier No. NBP01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Permanent Tattoo Pigment for NEO-9 Neonate Tattoo System

  • For proper tattooing and maintenance of tattoo system
  • Black colored permanent pigment
  • 0.5 oz. Bottle

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Capacity (English) 0.5 oz.
Capacity (Metric) 14.78 mL
Color Black
For Use With (Application) In-vivo and in-vitro Toxicology Assay
Type Permanent Tattoo Pigment
For Use With (Equipment) NEO-9 neonate tattoo system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.