Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIMS™ Skin Prep/Tissue Oil Applicator

Catalog No. 14370145
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-370-145 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-370-145 Supplier AIMS™ Supplier No. NSPA1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to apply skin prep/tissue oil to neonate rodent pups

  • Use to prevent tattoo fading
  • For applying skin prep solution to tail or paw of animal
  • Plastic handle
  • Hypoallergenic

 

Specifications

Color White
For Use With (Application) Applying skin prep solution to tail or paw of animal
Type Skin Preparation Applicator
Disposable Single-use
For Use With (Equipment) NEO-9 Neonate Tattoo System
Length (English) 3 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.