Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIMS™ Flex Arm LED Light for Stereotaxic Platforms

Catalog No. 11900100
Change view
Click to view available options
Certifications:
CE, UL, RoHS 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications
11-900-100 CE, UL, RoHS 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-900-100 Supplier AIMS™ Supplier No. ST13
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Attaches to any stereotaxic platform system to provide a bright LED light during procedures

    • Bright LED on 10 in. flex arm, no heat produced
    • Ultra strong magnetic base/puck system to anchor light on non-metallic surfaces
    • Use on any stereotaxic platform
    • Attaches directly to metallic surfaces
    • Universal wall plug power supply (110-220V) w/ 3-ft cord
    • Small base (1.625 in. diameter)

Specifications

Certifications CE, UL, RoHS 2
For Use With (Application) Stereotaxic Surgical Procedures
Type Stereotaxic LED Light
For Use With (Equipment) Use With Any Stereotaxic Equipment
Includes Magnetic Base w/Magnetic Puck
Length (English) 3 ft. Cord, 10 in. Flex Arm
Diameter (English) 1.625 in.
Diameter (Metric) 4.12 cm
Voltage Universal Input 110/220 VAC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.