Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine Laboratories AimScreen™ CLIA Waived Fentanyl Test Cassette, 1 ng/mL

Catalog No. 21124384
Change view
Click to view available options
Quantity:
25 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-124-384 25 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-124-384 Supplier Germaine Laboratories Supplier No. 42338
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

AimScreen™ CLIA Waived Fentanyl Test Cassette for over-the-counter and point-of-care use. For In Vitro Diagnostic Use. Results in 5 minutes.

  • For Over-the-Counter and Point-of-Care use
  • For In Vitro Diagnostic Use
  • Results in 5 minutes
  • Fentanyl Urine Test Cassette is an immunoassay intended for the qualitative detection of fentanyl in human urine at a cutoff concentration of 1 ng/mL
  • 25 individually foil wrapped tests per kit
  • The Fentanyl Urine Test Cassette is a rapid urine screening test that can be performed without the use of an instrument
  • The test utilizes a monoclonal antibody to selectively detect elevated levels of Fentanyl in urine
  • The Fentanyl Urine Test Cassette is an immunoassay based on the principle of competitive binding
  • The Fentanyl Urine Test Cassette yields a positive result when Fentanyl in urine exceeds 1 ng/mL

Specifications

CE Marker No
Clia Complexity Waived
Detectable Analytes Fentanyl (FEN)
Sensitivity 1 ng/mL
Type CLIA Waived Fentanyl Test Cassette
Disposable Single-use
Quantity 25 Tests/Kit
Test Time 5 min.
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.