Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimScreen™ DOA Negative Control

Catalog No. 23111361
Change view
Click to view available options
Quantity:
1 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-361 1 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-361 Supplier Germaine™ Laboratories Supplier No. 40003
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy

Germaine™ Laboratories AimScreen™ DOA Negative Controls monitor and validate the performance of DOA detection methods at levels established by SAMHSA, CAP/AACC and many state programs

  • Compatible with all quantitative and qualitative drug detection procedures which are sufficiently sensitive to detect the control constituents
  • Should be treated as any unknown specimen while following the specific protocol of the assay being used
  • Controls prepared from human based urine available as a negative
  • Cutoff is +3x to monitor the performance of drugs of abuse test devices
  • Each bottle contains stabilized human based urine
  • Human urine based matrix

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes Negative Control
For Use With (Equipment) All quantitative and qualitative drug detection procedures which are sufficiently sensitive to detect the control constituents.
Quantity 1 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.