Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimScreen™ Fentanyl (FEN) DipDevice

Catalog No. 23111376
Change view
Click to view available options
Quantity:
25 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-376 25 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-111-376 Supplier Germaine™ Laboratories Supplier No. 42335
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AimScreen™ FEN Urine Test is competitive binding, lateral flow immunochromatographic assays for qualitative and simultaneous detection of Fentanyl (FEN) 200 ng/mL in human urine

  • Rapidly screen for broad range of illicit and prescription drugs
  • Simple procedure: collect, dip and read-results in as little as five minutes
  • Test device remains in control of administrator
Note:  For forensic use only.

Specifications

Content And Storage 2°C to 30°C
Concentration 200 ng/mL
Detectable Analytes Fentanyl (FEN)
Sensitivity 200 ng/mL
Type Urine Test
For Use With (Application) For forensic use only; should not be used in a hospital lab.
Includes 25 tests/kit
Packaging Type Individually foil wrapped
Quantity 25 Tests/Kit
Test Time 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.