Learn More
Germaine Laboratories, Inc. AimScreen™ MultiDrug 9
Catalog No. 21010238
Change view
Click to view available options
Detectable Analytes:
Cocaine (COC)
Catalog No. Detectable Analytes
21-010-238 Cocaine (COC)
1 options
This item is not returnable. View return policy
Catalog No. 21-010-238 Supplier Germaine Laboratories, Inc. Supplier No. 40925
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed for the qualitative detection of drugs of abuse in urine
  • Rapidly screen for broad range of illicit and prescription drugs
  • AimScreen™ is a single-step immunoassay designed for the qualitative detection of drugs of abuse in urine
  • Rapidly screen for broad range of illicit and prescription drugs
  • 510(k) Cleared and CLIA-waived
  • Simple procedure collect, dip and read-results in as little as five minutes
  • Test device remains in control of administrator
Specifications
CE Marker No
Clia Complexity Waived
Detectable Analytes Cocaine (COC)
Type Dipdevice
Disposable Single-use
Test Time 5 min.
Form Ready-to-Use (RTU)
Operating Temperature Range 2°C to 30°C