Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimStep™ Cotinine DipDevice

Catalog No. 23111345
Change view
Click to view available options
Quantity:
1 Test
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-345 1 Test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-111-345 Supplier Germaine™ Laboratories Supplier No. 40010
Add to Cart
Add to Cart

Germaine™ Laboratories AimStep™ Cotinine DipDevice is a rapid, qualitative test for the detection of cotinine in urine

Designed to detect cotinine at a concentration of 200 ng/mL.

Specifications

Content And Storage 2°C to 30°C
Clia Complexity Moderate
Detectable Analytes Cotinine
Sample Type Urine
Type Urine DipDevice
Disposable Single-use
Quantity 1 Test
Test Time 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.